Learn More
Invitrogen™ eBioscience™ Protein Transport Inhibitor Cocktail (500X)
Suitable for use in assays in which prevention of protein transport to the extracellular space is desired.
Brand: Invitrogen™ 00-4980-03
59.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.09500kg Transport : UN number : 1170 Chem class : 3 Pack group : II
Description
- A cocktail of Brefeldin A and Monensin
- Suitable for use in assays in which prevention of protein transport to the extracellular space is desired
- It has been specifically formulated for use as a negative control for the Cell Stimulation Cocktail (plus protein transport inhibitors) (500X)
- Addition of this cocktail to cells in culture inhibits the intracellular protein secretory/transport pathway resulting in the accumulation of secreted proteins in the lumen of the endoplasmic reticulum and in the Golgi apparatus
- This accumulation can then be detected by intracellular staining and flow cytometric analysis or western blotting
Specifications
Protein Transport Inhibitor Cocktail | |
4 x 100 μL | |
eBioscience™ |
For Research Use Only. Not for use in diagnostic procedures.