missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Protein LoBind™ Tubes
Protein LoBind™ Tubes are specifically designed for use in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results.
£92.30 - £109.00
Specifications
Closure Color | Gray |
---|---|
Color | Colorless |
Disposable | Single-use |
No. per Pack | 200 |
Packaging | Bag |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15925180
|
Eppendorf™
0030122216 |
15 mL |
£92.30
Pack of 200 |
In Stock
Log in to see stock. |
|||||
15945180
|
Eppendorf™
0030122240 |
50 mL |
£109.00
Pack of 200 |
In Stock
Log in to see stock. |
|||||
Description
- Eppendorf™ LoBind™ material ensures optimized sample recovery for improved assay results
- Free of surface coating (e.g., silicone) to minimize the risk of sample interference
- Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
- Available in tube, microplate, and deepwell plate formats for easy-up scaling
- Precise lid sealing to minimize evaporation
Specifications
Gray | |
Single-use | |
Bag | |
PCR Clean | |
Screw Cap | |
Yes |
Colorless | |
200 | |
Autoclavable | |
Conical | |
Yes |