Thermo Scientific™ Protease Inhibitor Cocktails

Protect your sample from proteolytic degradation during cell lysis with these broad-spectrum protease inhibitor cocktails.

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: 78410

UNSPSC: 12352200

Code: PN

Additional Details:
Additional Details: Weight: 4.08230kg



Product Code. 11804101

Quantity Price
1 £ 166.0 / 2mL
EU Stock 9
Add to basket


Description and Specification

Specification

Formulation Proprietary components in DMSO
Sufficient For 200mL of sample
Format Liquid
Description Protease Inhibitor Cocktail
Composition Proprietary formulation
Quantity 2mL

Both ready-to-use formulations inhibit most serine, cysteine, and calpain proteases that cause general degradation of proteins in cell extracts. Metalloproteases are inhibited as well. The EDTA-free formulation is ideal for preparing samples that will be analyzed by 2D gel electrophoresis. The refrigerator-stable, 100X liquid solutions are easy to use: simply add the appropriate volume of cocktail to the sample (10μL per 1mL of cell extract) and continue with your procedure. The Protease Inhibitor can be used with animal cells and tissues, bacteria, plants and yeast.

Highlights:

  • Ready-to-use, 100X liquid cocktail
  • Available with EDTA (supplied in separate vial) or as an EDTA-free formulation
  • Effective for suppressing proteolytic activity in detergent-based cell lysis reagents
  • No loss of valuable freezer space; cocktail is stable at 4°C