missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Protease Inhibitor Cocktail V, EDTA-free

For inhibition of cysteine and serine proteases, but not not metalloproteases

Brand:  Alfa Aesar™ J65412.LQ

Code : NEW

Additional Details : Weight : 0.01000kg Transport : UN number : 3261 Chem class : 1a Pack group : III

 View more versions of this product

Product Code. 15496889

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

For inhibition of cysteine and serine proteases, but not not metalloproteases
Lyophilized Solid
−30° to −10°C
Hygroscopic
White
Protease Inhibitor Cocktail
1 Unit
Videos
SDS