Learn More
Thermo Scientific Chemicals Protease Inhibitor Cocktail I, Animal Free
Brand: Thermo Scientific Chemicals J65358.LQ
Code : NEW
Additional Details : Weight : 0.01000kg Transport : UN number : 3261 Chem class : 8 Pack group : III
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
White | |
Air Sensitive | |
Solid |
Protease Inhibitor Cocktail | |
-30°C to -10°C | |
1 Unit |
RUO – Research Use Only