missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Protease Inhibitor Cocktail, for general use

Brand:  Alfa Aesar™ J61852.XF

Code : NEW

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15424349

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

Liquid
2° to 8°C
Odorless
Protease Inhibitor Cocktail
1mL
Videos
SDS