missing translation for 'onlineSavingsMsg'
Learn More

PROSWIFT RP-4H, MONOLITH, 0.1MM x 250MM

Brand:  ThermoScientific BioLC Columns 164922

PROSWIFT RP-4H, MONOLITH, 0.1MM x 250MM

Code : NEW

Additional Details : Weight : 10.00000kg

Product Code. 15204848

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.