Learn More
R&D Systems™ Prostaglandin E2 Parameter Assay Kit, R&D Systems™
a 3.5 hour forward sequential competitive enzyme immunoassay designed to measure PGE2 in cell culture supernates, serum, plasma, and urine
Brand:  R&D Systems™ KGE004B
Code : Z1
Additional Details : CAS Number : 7732-18-5 Weight : 0.01000kg Transport : UN number : 2796 Chem class : 8 Pack group : II
 View more versions of this product
Product Code. 13488045
  • £359.05 / 1 set
Estimated Shipment
27-09-2018
Add to basket
Description
Description
This assay is based on the forward sequential competitive binding technique in which PGE2 present in a sample competes with horseradish peroxidase (HRP)-labeled PGE2 for a limited number of binding sites on a mouse monoclonal antibody. PGE2 in the sample is allowed to bind to the antibody in the first incubation. During the second incubation, HRP-labeled PGE2 binds to the remaining antibody sites. Following a wash to remove unbound materials, a substrate solution is added to the wells to determine the bound enzyme activity. The color development is stopped, and the absorbance is read at 450 nm. The intensity of the color is inversely proportional to the concentration of PGE2 in the sample.
Specifications
Specifications
Biotin
ELISA
1 kit
Multi-species
Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
ELISA kit
414pg/mL
1 x 96-well plate
Videos