missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Propylenediphosphonic acid, 98+%

Brand:  Alfa Aesar™ 030277.09

Code : NEW

Additional Details : CAS Number : 4671-82-3 Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 11307248

  • £334.10 / 10g


Estimated Shipment date
from Supplier 30-04-2018
Add to basket

Specifications

Chemical Identifiers

Propylenediphosphonic acid
4671-82-3
204.06
MFCD00050066

Specifications

Propylenediphosphonic acid
Powder
Odorless
MFCD00050066
10g
4671-82-3
≥98%
204.06
178°C
Videos
SDS