missing translation for 'onlineSavingsMsg'
Learn More

(±)-Propylene oxide, 99%, pure, Thermo Scientific Chemicals

Catalog Number p-188518
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
75-56-9
C3H6O
58.08
MFCD00005126
GOOHAUXETOMSMM-UHFFFAOYNA-N
propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane
6378
CHEBI:38685
2-methyloxirane
CC1CO1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
75-56-9
C3H6O
58.08
MFCD00005126
GOOHAUXETOMSMM-UHFFFAOYNA-N
propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane
6378
CHEBI:38685
2-methyloxirane
CC1CO1

(+/-)-Propylene oxide, 99%, C3H6O, CAS Number-75-56-9, 2,3-epoxypropane, epoxypropane, methyloxirane, propylene oxide, oxirane, methyl, propyleneoxide, 1,2-propylene oxide, propene oxide, propylene epoxide, 1,2-epoxypropane, 1L, 17, 6, 34.0 deg.C, CHEBI:38685, Undesignated, 0.8300g/mL, 200-879-2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -112.0°C
Density 0.8300g/mL
Boiling Point 34.0°C
Flash Point −37°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.3650 to 1.3670
Quantity 1 L
Beilstein 17, 6
Merck Index 15, 7969
Specific Gravity 0.83
Formula Weight 58.08
Vapor Pressure 590mbar at 20°C
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material (±)-Propylene oxide, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (+/-)-Propylene oxide
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 1
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'