missing translation for 'onlineSavingsMsg'
Learn More

Propylene Glycol, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5391329
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
This item is not returnable. View return policy
57-55-6
C3H8O2
76.095
DNIAPMSPPWPWGF-UHFFFAOYSA-N
1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost
1030
CHEBI:16997
propane-1,2-diol
CC(CO)O
This item is not returnable. View return policy
57-55-6
C3H8O2
76.095
DNIAPMSPPWPWGF-UHFFFAOYSA-N
1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost
1030
CHEBI:16997
propane-1,2-diol
CC(CO)O

High quality chemicals for laboratory and specialized industrial use

Quantity 500 mL
Formula Weight 76.1
Flash Point 99.00°C
Grade BAKER ANALYZED™ ACS Reagent
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.04kg
Chemical Name or Material Propylene Glycol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'