missing translation for 'onlineSavingsMsg'
Learn More

Propylene carbonate, 99%, Thermo Scientific Chemicals

Catalog Number p-3346462
Click to view available options
Quantity:
250 g
1000 g
2.5 kg
5 kg
This item is not returnable. View return policy
108-32-7
C4H6O3
102.09
MFCD00005385,MFCD00798264,MFCD00798265
RUOJZAUFBMNUDX-UHFFFAOYNA-N
propylene carbonate, 1,2-propylene carbonate, 1,2-propanediol cyclic carbonate, texacar pc, 1,3-dioxolan-2-one, 4-methyl, cyclic propylene carbonate, arconate 5000, 1,2-propanediol carbonate, 1-methylethylene carbonate, cyclic 1,2-propylene carbonate
7924
4-methyl-1,3-dioxolan-2-one
CC1COC(=O)O1
This item is not returnable. View return policy
108-32-7
C4H6O3
102.09
MFCD00005385,MFCD00798264,MFCD00798265
RUOJZAUFBMNUDX-UHFFFAOYNA-N
propylene carbonate, 1,2-propylene carbonate, 1,2-propanediol cyclic carbonate, texacar pc, 1,3-dioxolan-2-one, 4-methyl, cyclic propylene carbonate, arconate 5000, 1,2-propanediol carbonate, 1-methylethylene carbonate, cyclic 1,2-propylene carbonate
7924
4-methyl-1,3-dioxolan-2-one
CC1COC(=O)O1

Propylene carbonate is a polar, aprotic solvent used in paints and coatings and as a high-permittivity component of electrolytes in lithium batteries. It plays an important role in adhesives, paint strippers, cosmetics, plasticizer, hard surface cleaner, resin cleaner, fiberglass, polyester and polyurethane cleaner. Furthermore, it acts as a cleaner in polyurethane, a carburetor and is used in stereo lithography parts. In electrospray ionization mass spectrometry, it is doped into low surface tension solutions to enhance analyte charging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propylene carbonate is a polar, aprotic solvent used in paints and coatings and as a high-permittivity component of electrolytes in lithium batteries. It plays an important role in adhesives, paint strippers, cosmetics, plasticizer, hard surface cleaner, resin cleaner, fiberglass, polyester and polyurethane cleaner. Furthermore, it acts as a cleaner in polyurethane, a carburetor and is used in stereo lithography parts. In electrospray ionization mass spectrometry, it is doped into low surface tension solutions to enhance analyte charging.

Solubility
Miscible with water, alcohol, ether, acetone, benzene, chloroform and ethyl acetate.

Notes
Moisture sensitive. Hygroscopic. Incompatible with acids and bases.
TRUSTED_SUSTAINABILITY
Melting Point °C to 49°C
Density 1.204
Boiling Point 240°C to 243°C
Flash Point 132°C (270°F)
Assay Percent Range 99%
Refractive Index 1.421
Quantity 250 g
Solubility Information Miscible with water,alcohol,ether,acetone,benzene,chloroform and ethyl acetate.
Formula Weight 102.09
Physical Form Liquid
Chemical Name or Material Propylene Carbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'