missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propylene carbonate, 99.5%, anhydrous, AcroSeal™, Thermo Scientific Chemicals
17.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -49.0°C |
Density | 1.2040g/mL |
Boiling Point | 240.0°C to 243.0°C |
Flash Point | 116°C |
Assay Percent Range | 99.4% min. (GC) |
Packaging | AcroSeal™ Glass Bottle |
Refractive Index | 1.4200 to 1.4220 |
Quantity | 100 mL |
Specific Gravity | 1.204 |
Solubility Information | Solubility in water: 200g/L (25°C). Other solubilities: miscible with acetone,benzene,chloroform,dmf,,ether,ethyl acetate and ethyl alcohol,slightly soluble in carbon tetrachloride,soluble in most organic solvents |
Show More |
Safety and Handling
Product Identifier
- Propylene carbonate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'