missing translation for 'onlineSavingsMsg'
Learn More

Propylbenzene, 98%, Thermo Scientific Chemicals

Catalog Number 10558853
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
103-65-1
C9H12
120.20
MFCD00009377
ODLMAHJVESYWTB-UHFFFAOYSA-N
n-propylbenzene, 1-phenylpropane, phenylpropane, benzene, propyl, 1-propylbenzene, isocumene, n-propyl benzene, propyl-benzene, unii-0wr86zhg2z, propylbenzene, n
7668
CHEBI:42630
propylbenzene
CCCC1=CC=CC=C1
This item is not returnable. View return policy
103-65-1
C9H12
120.20
MFCD00009377
ODLMAHJVESYWTB-UHFFFAOYSA-N
n-propylbenzene, 1-phenylpropane, phenylpropane, benzene, propyl, 1-propylbenzene, isocumene, n-propyl benzene, propyl-benzene, unii-0wr86zhg2z, propylbenzene, n
7668
CHEBI:42630
propylbenzene
CCCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -99°C
Color Yellow
Density 0.8600g/mL
Boiling Point 158°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2CH2CH3
Refractive Index 1.4900 to 1.493
Quantity 500 g
Beilstein 05, 390
Merck Index 15, 7957
Specific Gravity 0.86
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: miscible with ethanol,ether and acetone
Formula Weight 120.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Propylbenzene
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Propylbenzene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'