missing translation for 'onlineSavingsMsg'
Learn More

Propylamine, 98%, pure, Thermo Scientific Chemicals

Catalog Number 10500522
Click to view available options
Quantity:
5 mL
1 L
5 L
Packaging:
Glass Bottle
Plastic drum
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
107-10-8
MFCD00008205
WGYKZJWCGVVSQN-UHFFFAOYSA-N
propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine
7852
CHEBI:39870
propan-1-amine
CCCN
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
107-10-8
MFCD00008205
WGYKZJWCGVVSQN-UHFFFAOYSA-N
propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine
7852
CHEBI:39870
propan-1-amine
CCCN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -83.0°C
Color Colorless to Yellow
Density 0.7190g/mL
Boiling Point 48.0°C
Flash Point −30°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C3H9N
Linear Formula CH3CH2CH2NH2
Refractive Index 1.3870 to 1.3890
Quantity 5 mL
Specific Gravity 0.719
Molecular Weight (g/mol) 59.112
Formula Weight 59.11
Percent Purity 98%
Grade Pure
Physical Form Liquid
Chemical Name or Material Propylamine, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'