missing translation for 'onlineSavingsMsg'
Learn More

Propionic acid, sodium salt, 99.0-100.5%, Thermo Scientific Chemicals

Catalog Number p-3758710
11.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
137-40-6
C3H5NaO2
96.06
MFCD00002759
JXKPEJDQGNYQSM-UHFFFAOYSA-M
sodium propionate, sodium propanoate, propionic acid sodium salt, propanoic acid, sodium salt, napropion, ocuseptine, deketon, impedex, keenate, propiofar
2723816
sodium;propanoate
CCC(=O)[O-].[Na+]
This item is not returnable. View return policy
137-40-6
C3H5NaO2
96.06
MFCD00002759
JXKPEJDQGNYQSM-UHFFFAOYSA-M
sodium propionate, sodium propanoate, propionic acid sodium salt, propanoic acid, sodium salt, napropion, ocuseptine, deketon, impedex, keenate, propiofar
2723816
sodium;propanoate
CCC(=O)[O-].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 285°C to 286°C
Color White
Flash Point >250°C
Infrared Spectrum Authentic
Assay Percent Range 99.0-100.5%
Packaging Plastic bottle
Linear Formula C2H5CO2Na
Quantity 25 g
Beilstein 02, 234
Merck Index 15, 8798
Solubility Information Solubility in water: very soluble. Other solubilities: soluble in alcohol
Formula Weight 96.06
Percent Purity 99.0-100.5%
Physical Form Crystalline Powder
Chemical Name or Material Propionic acid, sodium salt
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'