missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propionic acid, ACS reagent, Thermo Scientific Chemicals
12.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 mL
500 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -22.0°C |
Density | 0.9900g/mL |
Boiling Point | 141.0°C |
Flash Point | 51°C |
Packaging | Glass bottle |
Linear Formula | C2H5CO2H |
Refractive Index | 1.3850 to 1.3870 |
Quantity | 100 mL |
Merck Index | 15,7939 |
Specific Gravity | 0.99 |
Show More |
Safety and Handling
Product Identifier
- Propionic acid
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : propionic acid
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'