missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Propionamide, 98%

Brand:  Alfa Aesar™ A10521.36

Code : NEW

Additional Details : CAS Number : 79-05-0 Weight : 0.50000kg

Quantity 500g

 View more versions of this product

Product Code. 11488783

  • £402.80 / 500g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Description

Description

Applications
Propionamide was used as adsorbent in the determination of adsorption isotherms of acetamide and propionamide on multi-wall carbon nanotube1. It was used in a robust screening method to study biotransformations using (+)-γ-lactamase enzyme. Propionamide on γ-irradiation reacts with sulfur dioxide and this reaction has been studied by ESR spectroscopy, gas absorption measurements and X-ray diffraction3

Solubility
Soluble in water, chloroform, ethanol, diethyl ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids, bases, strong reducing agents.
Specifications

Chemical Identifiers

Propionamide
CH3CH2CONH2
MFCD00008039
79-05-0
73.1
14,7824

Specifications

Propionamide
98%
73.1
14,7824
76° to 79°C
79-05-0
CH3CH2CONH2
MFCD00008039
213°C
500g
Videos
SDS