missing translation for 'onlineSavingsMsg'
Learn More

Propionamide, 97%, Thermo Scientific Chemicals

Catalog Number 10701935
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
79-05-0
C3H7NO
73.09
MFCD00008039
QLNJFJADRCOGBJ-UHFFFAOYSA-N
propionamide, propylamide, propionic amide, propionimidic acid, propanimidic acid, propionic acid amide, amid kyseliny propionove, propanimidic acid van, propionimidic acid van, unii-qk07g0hp47
6578
CHEBI:45422
propanamide
CCC(=O)N
This item is not returnable. View return policy
79-05-0
C3H7NO
73.09
MFCD00008039
QLNJFJADRCOGBJ-UHFFFAOYSA-N
propionamide, propylamide, propionic amide, propionimidic acid, propanimidic acid, propionic acid amide, amid kyseliny propionove, propanimidic acid van, propionimidic acid van, unii-qk07g0hp47
6578
CHEBI:45422
propanamide
CCC(=O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 79.0°C to 83.0°C
Color White to Yellow
Boiling Point 213.0°C
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula C2H5CONH2
Quantity 25 g
Beilstein 02, 243
Merck Index 15, 7938
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in alcohol, ether, chloroform
Formula Weight 73.09
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Propionamide, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Propionamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'