missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propiolic acid, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Propiolic acid is a reagent used in the preparation of transition metal complexes, haloalkyl propiolates and halopropenoates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Propiolic acid is a reagent used in the preparation of transition metal complexes, haloalkyl propiolates and halopropenoates.
Solubility
Miscible with water, ether, ethanol and chloroform.
Notes
Store in cool place. Incompatible with bases, oxidizing agents and reducing agents.
Propiolic acid is a reagent used in the preparation of transition metal complexes, haloalkyl propiolates and halopropenoates.
Solubility
Miscible with water, ether, ethanol and chloroform.
Notes
Store in cool place. Incompatible with bases, oxidizing agents and reducing agents.
Specifications
Melting Point | 18°C |
Density | 1.136 |
Boiling Point | 102°C (200 mmHg) |
Flash Point | 58°C (136°F) |
Refractive Index | 1.433 |
Quantity | 5 g |
UN Number | UN2920 |
Beilstein | 878176 |
Merck Index | 14,7824 |
Solubility Information | Miscible with water,ether,ethanol and chloroform. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'