missing translation for 'onlineSavingsMsg'
Learn More

Propargylamine hydrochloride, 95%, Thermo Scientific Chemicals

Catalog Number p-7059016
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
15430-52-1
C3H6ClN
91.54
MFCD00012907
IKXNIQJDNKPPCH-UHFFFAOYSA-N
propargylamine hydrochloride, prop-2-yn-1-amine hydrochloride, 2-propynylamine hydrochloride, propargylamine hcl, 2-propynylamine hcl, 3-amino-1-propyne hydrochloride, prop-2-ynylamine hydrochloride, 2-propyn-1-amine, hydrochloride, pubchem18845, acmc-1bvqh
11205720
prop-2-yn-1-amine;hydrochloride
[H+].[Cl-].NCC#C
This item is not returnable. View return policy
15430-52-1
C3H6ClN
91.54
MFCD00012907
IKXNIQJDNKPPCH-UHFFFAOYSA-N
propargylamine hydrochloride, prop-2-yn-1-amine hydrochloride, 2-propynylamine hydrochloride, propargylamine hcl, 2-propynylamine hcl, 3-amino-1-propyne hydrochloride, prop-2-ynylamine hydrochloride, 2-propyn-1-amine, hydrochloride, pubchem18845, acmc-1bvqh
11205720
prop-2-yn-1-amine;hydrochloride
[H+].[Cl-].NCC#C

Propargylamine hydrochloride is a useful intermediates in the preparation of drugs. It is used in pharmaceutical, industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 179°C to 182°C
Quantity 1 g
UN Number UN2811
Sensitivity Hygroscopic
Solubility Information Soluble in water (partly),methanol,and ethanol.
Formula Weight 91.54
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Propargylamine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'