missing translation for 'onlineSavingsMsg'
Learn More

Propargyl alcohol, 99%, Thermo Scientific Chemicals

Catalog Number 10523301
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
107-19-7, 50-00-0
C3H4O
56.06
MFCD00002912
TVDSBUOJIPERQY-UHFFFAOYSA-N
propargyl alcohol, 2-propyn-1-ol, ethynylcarbinol, 1-propyn-3-ol, propynyl alcohol, 2-propynyl alcohol, 2-propynol, 3-propynol, 1-hydroxy-2-propyne, 3-hydroxy-1-propyne
7859
CHEBI:28905
prop-2-yn-1-ol
OCC#C
This item is not returnable. View return policy
107-19-7, 50-00-0
C3H4O
56.06
MFCD00002912
TVDSBUOJIPERQY-UHFFFAOYSA-N
propargyl alcohol, 2-propyn-1-ol, ethynylcarbinol, 1-propyn-3-ol, propynyl alcohol, 2-propynyl alcohol, 2-propynol, 3-propynol, 1-hydroxy-2-propyne, 3-hydroxy-1-propyne
7859
CHEBI:28905
prop-2-yn-1-ol
OCC#C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -53.0°C
Color Colorless to Yellow
Density 0.9490g/mL
Boiling Point 114.0°C to 115.0°C
Flash Point 34°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula HC≡CCH2OH
Refractive Index 1.4300 to 1.4330
Quantity 250 mL
Beilstein 01,454
Fieser 09,394
Merck Index 15,7923
Specific Gravity 0.949
Solubility Information Solubility in water: miscible. Other solubilities: miscible with benzene, chloroform, ethanol, ether,, 1,2-dichloroethane, acetone, dioxane, thf,pyridine, moderately soluble in ccl4, immiscible with aliphatic hydrocarbons
Viscosity 1.58 mPa.s (20°C)
Formula Weight 56.06
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Propargyl alcohol, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Propargyl alcohol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H350-May cause cancer.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'