Learn More
Propan-1-ol, Extra Pure, SLR, Fisher Chemical™
Brand: Fisher Chemical P/7480/25
Code : 21
Additional Details : CAS Number : 71-23-8 Weight : 20.07500kg Transport : UN number : 1274 Chem class : 3 Pack group : II
Packaging | Metal can |
---|---|
Quantity | 25L |
Chemical Identifiers
71-23-8 | |
60.10 | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
1031 | |
propan-1-ol |
C3H8O | |
MFCD00002941 | |
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
CHEBI:28831 | |
CCCO |
Specifications
71-23-8 | |
MFCD00002941 | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
propan-1-ol | |
1031 | |
60.1g/mol | |
97°C | |
-127°C | |
25L | |
2.07 | |
2.2 mPaS at 20°C |
C3H8O | |
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
CCCO | |
60.10 | |
CHEBI:28831 | |
Liquid | |
Colorless | |
7 | |
Metal can | |
25mbar at 20°C |