Learn More
Propan-1-ol, Certified AR for Analysis, Fisher Chemical™
C3H8O, CAS Number-71-23-8, 1-hydroxypropane, optal, propanol, osmosol extra, n-propyl alcohol, n-propanol, ethylcarbinol, 1-propanol, propyl alcohol, propylic alcohol, 2.5L, 97 deg.C, CHEBI:28831, Colorless, 60.1g/mol, BDERNNFJNOPAEC-UHFFFAOYSA-N, propan-1-ol, -127 deg.C, 2941, 60.096
Brand: Fisher Chemical P/7481/15
20.46 GBP valid until 2022-06-30
Use promo code "16496" to get your promotional price.
Code : 22
Additional Details : CAS Number : 71-23-8 Weight : 0.80300kg Transport : UN number : 1274 Chem class : 3 Pack group : II
Packaging | Amber glass bottle |
---|---|
Quantity | 1L |
Description
C3H8O, CAS Number-71-23-8, 1-hydroxypropane, optal, propanol, osmosol extra, n-propyl alcohol, n-propanol, ethylcarbinol, 1-propanol, propyl alcohol, propylic alcohol, 2.5L, 97 deg.C, CHEBI:28831, Colorless, 60.1g/mol, BDERNNFJNOPAEC-UHFFFAOYSA-N, propan-1-ol, -127 deg.C, 2941, 60.096
Chemical Identifiers
71-23-8 | |
60.10 | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
1031 | |
propan-1-ol |
C3H8O | |
MFCD00002941 | |
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
CHEBI:28831 | |
CCCO |
Specifications
71-23-8 | |
MFCD00002941 | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
propan-1-ol | |
1031 | |
60.1g/mol | |
97°C | |
-127°C | |
1L | |
2.07 | |
2.2 mPaS at 20°C |
C3H8O | |
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
CCCO | |
60.10 | |
CHEBI:28831 | |
Liquid | |
Colorless | |
7 | |
Amber glass bottle | |
25mbar at 20°C |
Safety and Handling
- 1-Propanol
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.