missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ProPac™ IMAC-10 LC Columns

Resolve target proteins using a single column in a high-resolution gradient run with ProPac IMAC-10 LC Columns.

Brand:  Thermo Scientific™ 063710

Code : 16

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 11311873

  • £288.00 / Each


Estimated Shipment
10-05-2018
Add to basket

Description

Description

Tailored Specificity for IMAC Separation of Proteins
  • Retention control by imidazole concentration or pH gradient
  • Stable between pH 2 to 12, compatible with common protein purification reagents (nonionic detergents, reducing agents)
  • Effective under native or denaturing HPLC conditions
  • Wide range of metal-specific applications, including the separation of His-tagged protein aggregation variants, prion peptide analysis and separation of monoclonal antibodies
  • Shipped free from any metal and ready to be charged with the metal of your choice.
  • Reusable columns with metal tailored specificity
  • Resolve target proteins using a single column in a high resolution gradient run
  • High purity separations of metal-binding proteins
Includes: 10µm, nonporous, polymeric beads coated with a hydrophilic layer, then grafted with poly(IDA) chains.
Specifications

Specifications

Loading Column
9mm
ProPac IMAC-10 Column
3000psi
10μm
Nonporous
Loading Column
Affinity
0.1 to 10mL/min.
50mm
Polymer, Spherical Nonporous
2 to 12
60°C
Poly (IDA) grafts
Videos