missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proliferin-related Protein/Plfr Rat anti-Mouse, Clone: 219310, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | Proliferin-related Protein/Plfr |
---|---|
Clone | 219310 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Proliferin-related Protein/Plfr Glu26-Phe244 Accession # P04769 |
Gene ID (Entrez) | 5617 |
Gene Accession No. | P04769 |
Description
Proliferin-related Protein/Plfr Monoclonal specifically detects Proliferin-related Protein/Plfr in Mouse samples. It is validated for Western Blot.Specifications
Proliferin-related Protein/Plfr | |
Mouse myeloma cell line NS0-derived recombinant mouse Proliferin-related Protein/Plfr Glu26-Phe244 Accession # P04769 | |
P04769 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Rat | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
219310 | |
5617 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects mouse Proliferin-related Protein/Plfr in direct ELISAs and Western blots. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only