missing translation for 'onlineSavingsMsg'
Learn More

Progesterone Receptor, eFluor 660, clone: KMC912, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-9764-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15510127

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »

Disclaimers:

For Research Use Only.


Description

Description

This KMC912 monoclonal antibody reacts with human progesterone receptor (PgR, PR), a member of a superfamily of nuclear receptors that are ligand-dependent transcriptional regulators. The human PgR exists in alpha and beta forms, 94kDa and 120kDa respectively. In most human cells, the alpha and beta forms are expressed at similar levels and predominately form heterodimers. Progestin binding to PgR causes a conformational change, allowing dissociation of bound chaperone proteins and subsequent dimerization with either PgRa or PgRb. Following activation, dimerized PgR can directly bind to DNA through progestin response elements (PRE) leading to chromatin remodeling and subsequent downregulation or transcription of the target gene.The PgR plays a key role in controlling gene expression in breast, uterine, brain, and cardiovascular tissue during development. The presence of the PgR in breast tissue is indicative of improved survival and a better response to endocrine therapy. In breast and endometrial cancer progression, a predominance of either the alpha or beta form occurs, suggesting disregulation in the PgRa:PgRb ratio is an early event in cancer. In cases of ductal carcinoma in situ and invasive ductal carcinoma, there is predominance of the alpha form while in uterine cancer a loss of either form is common.
Specifications

Specifications

Progesterone Receptor
KMC912
eFluor 660
Conjugated
Murine
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Microscopy
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
PR, PgR
IgG1
RUO
Primary
Human
Videos