missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Profood™ 87-315 Series Blue Natural Rubber Latex Gloves

Reusable latex gloves chlorinated outside to increase mechanical and chemical resistance.

Brand:  Ansell Edmont™ 87315070

Code : NEW

Additional Details : Weight : 0.70000kg

 View more versions of this product

Product Code. 11756744

  • £15.47 / 12 pairs


Estimated Shipment
25-05-2018
Add to basket

Description

Description

  • Made from 100% pure latex

Applications:

  • Poultry processing
  • Beef and pig processing
  • Fish processing
  • Fruit and vegetable processing
Specifications

Specifications

Natural Rubber Latex
Natural Rubber Latex
Cotton Flocking
87-315
144 Pairs
Gloves
Natural Rubber Latex
Pinked cuff
Liquid and mechanical resistant
87-315
0.4mm
Blue
Videos