missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Profood™ 87-305 Series Blue Natural Rubber Latex Gloves

Reusable latex gloves chlorinated inside to minimise the risk of allergy.

Brand:  Ansell Edmont™ 87305070

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12647021

  • £20.80 / 12 pairs


Estimated Shipment
15-02-2018
Add to basket

Description

Description

  • Made from 100% pure latex

Applications:

  • Poultry processing
  • Beef and pig processing
  • Fish processing
  • Fruit and vegetable processing
Specifications

Specifications

Natural Rubber Latex
Natural Rubber Latex
Unflocked
87-305
144 Pairs
Gloves
Natural Rubber Latex
Pinked cuff
Liquid and mechanical resistant
87-305
0.4mm
Blue
Videos