missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell Edmont™ Profil™ Plus 87-850 Series Yellow Natural Rubber Latex Immersion Gloves
Click to view available options
Size:
8
9
Description
- Specially treated to reduce the risk of allergy
- Excellent resistance to mechanical hazards and enhanced chemical resistance
- 100% cotton flocklining - soft, comfortable and absorbent
- Ideal for janitorial use and handling in wet conditions
Specifications
Back Material | Rubber Latex |
Cuff Material | Natural Rubber Latex |
Coating Material | Natural Rubber Latex |
Latex Free | No |
Product Type | Immersion Gloves |
Style | 87-850 |
Thickness (Metric) | 0.55 mm |
Glove Cuff Color | Yellow |
Glove Cuff Style | Pinked |
Liner Material | Cotton Flocking |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'