missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ ProCulture Warming Plate, Ambient to 37°C, Aluminum
Shape: Rectangle; Size: 7 x 8 in.
Brand: Bel-Art™ F37015-1000
Code : NEW
Additional Details : Weight : 1.31400kg
Description
Shape: Rectangle; Size: 7 x 8 in.
- Increase cell viability during cell passaging with this highly accurate warming plate that keeps cells near an optimal 37°C (±1°C) outside of the incubator.
- Avoids numerous entries into the CO2 incubator which alter its atmosphere, harming ongoing experiments in the incubated cultures
- Reduces cell stress and allows enzymes like trypsin to work efficiently
- Can be used to maintain the warmth of reagents inside the hood
- Fits easily in cell culture hoods or on the workbench
- Temperature remains uniform across the entire anodized aluminum warming plate; measures 17.8 D x 20.3 cmW (7 x 8 in.)
- 15-minute count down timer keeps track of time sensitive applications
- UV resistant polypropylene housing will not break down under germicidal UV lights
- Water resistant; safe for cleaning with Ethanol or IPA
- 220V is CE marked
Compliance
UL approval pending.
Specifications
Warming Plate | |
7 in. x 8 in. | |
220 V | |
7 x 8 in. | |
10 in. | |
2.25 in. | |
5.7 cm | |
22 w |
CE | |
Rectangle | |
Aluminum | |
Ambient to 37°C | |
25.4 cm | |
9.75 in. | |
24.9 cm |