missing translation for 'onlineSavingsMsg'
Learn More

Procaterol hydrochloride, Tocris Bioscience™

Potent β2 agonist

Brand:  Tocris (Bio-Techne) 110210

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13438493

  • £135.24 / 10mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specific and very potent β2 agonist.
Specifications

Chemical Identifiers

62929-91-3
326.821
procaterol hcl, procaterol hydrochloride hemihydrate, 5-1-hydroxy-2-isopropylamino butyl-8-hydroxycarbostyril hydrochloride, r*,s*-1-8-hydroxy-5-1-hydroxy-2-isopropylamino butyl quinolin-2 1h-one monohydrochloride, 2 1h-quinolinone, 8-hydroxy-5-1-hydroxy-2-1-methylethyl amino butyl-, monohydrochloride, r*,s*-+-, unii-4vd1brt7t8 component, 8-hydroxy-5-1s,2r-1-hydroxy-2-isopropylamino butyl-1h-quinolin-2-one hydrochloride, r*,s*-+/--8-hydroxy-5-1-hydroxy-2-1-methylethyl amino butyl-2 1h-quinolinone monohydrochloride, hydrate, rel-8-hydroxy-5-1r,2s-1-hydroxy-2-1-methylethyl amino butyl-2 1h-quinolinone hydrochloride, hydrate
8-hydroxy-5-[(1S,2R)-1-hydroxy-2-(propan-2-ylamino)butyl]-1H-quinolin-2-one;hydrochloride
C16H23ClN2O3
AEQDBKHAAWUCMT-KKJWGQAZSA-N
16219912
CCC(C(C1=C2C=CC(=O)NC2=C(C=C1)O)O)NC(C)C.Cl

Specifications

10mg
62929-91-3
procaterol hcl, procaterol hydrochloride hemihydrate, 5-1-hydroxy-2-isopropylamino butyl-8-hydroxycarbostyril hydrochloride, r*,s*-1-8-hydroxy-5-1-hydroxy-2-isopropylamino butyl quinolin-2 1h-one monohydrochloride, 2 1h-quinolinone, 8-hydroxy-5-1-hydroxy-2-1-methylethyl amino butyl-, monohydrochloride, r*,s*-+-, unii-4vd1brt7t8 component, 8-hydroxy-5-1s,2r-1-hydroxy-2-isopropylamino butyl-1h-quinolin-2-one hydrochloride, r*,s*-+/--8-hydroxy-5-1-hydroxy-2-1-methylethyl amino butyl-2 1h-quinolinone monohydrochloride, hydrate, rel-8-hydroxy-5-1r,2s-1-hydroxy-2-1-methylethyl amino butyl-2 1h-quinolinone hydrochloride, hydrate
CCC(C(C1=C2C=CC(=O)NC2=C(C=C1)O)O)NC(C)C.Cl
326.821
326.82
Procaterol hydrochloride
C16H23ClN2O3
AEQDBKHAAWUCMT-KKJWGQAZSA-N
8-hydroxy-5-[(1S,2R)-1-hydroxy-2-(propan-2-ylamino)butyl]-1H-quinolin-2-one;hydrochloride
16219912
>99%
Videos
Documents