missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Procaspase-3 Activator

Brand:  Alfa Aesar™ J64824.MF

Code : NEW

Additional Details : CAS Number : 315183-21-2 Weight : 0.00050kg Transport : UN number : 3077 Chem class : 1 Pack group : III

 View more versions of this product

Product Code. 15466599

  • £244.65 / 50mg


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Specifications

Powder
Protease
50mg
315183-21-2
PAC-1; (4-Benzylpiperazino)acetic acid-(3-allyl-2-hydroxybenzylidene)hydrazide
Yellow
392.49g/mol
−30° to −10°C
C23H28N4O2
Light Sensitive
Videos
SDS