missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ProcartaPlex™ 96 well Flat Bottom Plate, black
Ideal for use with all ProcartaPlex Luminex Assys.The plate is included in all ProcartaPlex Multiplex and Basic Kits
Brand: Invitrogen™ EPX-44444-000
Code : NEW
Additional Details : Weight : 0.47500kg
Description
Reactivity/Species: Canine, Human, Mouse, Non-human primate, Pig, RatSpecifications
Fluorescence | |
96-well Microplate | |
ProcartaPlex™ |