missing translation for 'onlineSavingsMsg'
Learn More

Procainamide hydrochloride, 99%, Thermo Scientific Chemicals

Catalog Number 10733211
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
614-39-1
C13H21N3O·HCl
271.78
MFCD00012998
ABTXGJFUQRCPNH-UHFFFAOYSA-N
procainamide hydrochloride, procainamide hcl, pronestyl, procanbid, procapan, procan, procainhydrochlorid, procan sr, procainii chloridum, promide hydrochloride
66068
CHEBI:8429
4-amino-N-[2-(diethylamino)ethyl]benzamide;hydrochloride
CCN(CC)CCNC(=O)C1=CC=C(C=C1)N.Cl
614-39-1
C13H21N3O·HCl
271.78
MFCD00012998
ABTXGJFUQRCPNH-UHFFFAOYSA-N
procainamide hydrochloride, procainamide hcl, pronestyl, procanbid, procapan, procan, procainhydrochlorid, procan sr, procainii chloridum, promide hydrochloride
66068
CHEBI:8429
4-amino-N-[2-(diethylamino)ethyl]benzamide;hydrochloride
CCN(CC)CCNC(=O)C1=CC=C(C=C1)N.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 166°C to 170°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2NC6H4CONHCH2CH2N(C2H5)2·HCl
Quantity 25 g
Beilstein 14, III, 1077
Merck Index 15, 7874
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,slightly soluble in chloroform,very sparingly soluble in benzene and ether
Formula Weight 271.78
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Procainamide hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Procainamide hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'