missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Pro-MMP-9 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 4 publications
POA
Specifications
Antigen | MMP-9 |
---|---|
Clone | 116134 |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Classification | Monoclonal |
Description
MMP-9 Monoclonal specifically detects MMP-9 in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
MMP-9 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP9, MMP-9, type V collagenase | |
MMP9 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
116134 | |
ELISA | |
Unconjugated | |
Rat | |
Mouse | |
P41245.1 | |
4318 | |
Mouse myeloma cell line NS0-derived recombinant mouse MMP-9 Ala20-Pro730 Accession # P41245.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Pro-MMP-9 in ELISAs. This antibody does not recognize the mature form of Pro-MMP-9. |
For Research Use Only