Pro-Lab Diagnostics Microbank™ Bacterial and Fungal Preservation System

Designed to simplify the storage and retrieval of bacterial and fungal cultures.  Prolab Diagnostics™ Microbank™ Bacterial and Fungal Preservation System is a ready-to-use system.

Overview
Brand: Pro-Lab Diagnostics

Manufacturer Part Number: PL.170Y

UNSPSC: 41104100

Code: LE

Additional Details:
Additional Details: Weight: 0.80000kg



Product Code. 11907037

Quantity Price
1 £ 101.0 / Pack of 80
In Stock 8
Add to basket


Description and Specification

Specification

Storage Requirements -70°C
Product Type Cryogenic Storage Vial
Closure Color Yellow
Quantity 80 Pack
For Use With (Application) Designed to simplify the storage and retrieval of bacterial and fungal cultures

  • Incorporates treated beads and a special cryopreservative solution
  • Each vial contains 25 beads, providing repeated identical cultures from the original organism
  • Specially formulated preservative ensures longer survival of fastidious cultures and higher quantitative recoveries
  • 80 Vials supplied in a freezer box with“see through”cover
  • Available in 5 cap/bead colors or mixed (16 vials each of blue, green, light blue, red, and yellow)