Learn More
Pro-Lab Diagnostics Microbank™ Bacterial and Fungal Preservation System
Designed to simplify the storage and retrieval of bacterial and fungal cultures.  Prolab Diagnostics™ Microbank™ Bacterial and Fungal Preservation System is a ready-to-use system.
Brand:  Pro-Lab Diagnostics PL.170LB
UNSPSC : 41104100
Code : LE
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11947037
  • £97.30 / Pack of 80
In Stock 7
Add to basket
Description
Description
  • Incorporates treated beads and a special cryopreservative solution
  • Each vial contains 25 beads, providing repeated identical cultures from the original organism
  • Specially formulated preservative ensures longer survival of fastidious cultures and higher quantitative recoveries
  • 80 Vials supplied in a freezer box with“see through”cover
  • Available in 5 cap/bead colors or mixed (16 vials each of blue, green, light blue, red, and yellow)
Specifications
Specifications
Light Blue
80 Pack
Designed to simplify the storage and retrieval of bacterial and fungal cultures
Cryogenic Storage Vial
-70°C