missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Pro-HGF Activator Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | HGF Activator |
---|---|
Clone | 198811 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
HGF Activator Monoclonal specifically detects HGF Activator in Mouse samples. It is validated for Western Blot.Specifications
HGF Activator | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q9R098 | |
3083 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Pro-HGF Activator Gln35-Ser653 (predicted) Accession # Q9R098 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Pro-HGF Activator in Western blots. In Western blots, only the HGFA precursor is recognized. In Western blots, no cross-reactivity with the active form of recombinant mouse HGFA, recombinant human (rh) HGFA, or rhKallikreins 3, 5, and 11 is observed. |
198811 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HGF activator, HGFA, Hgfac | |
HGFAC | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only