missing translation for 'onlineSavingsMsg'
Learn More

Pro EGF Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4289

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438654

  • £362.60 / 100µg

Estimated Shipment
17-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Pro EGF Polyclonal antibody AF4289 Specifically detects Pro EGF in Human samples. It is validated for Western Blotting
Specifications

Specifications

EGF
LYOPH
Western Blot 0.1 ug/mL
CAA28240
beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), HOMG4, pro-epidermal growth factor, URG, Urogastrone
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Pro-EGF in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse Pro-EGF is observed and less than 1% cross-reactivity with mature recombinant human EGF is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
EGF
Mouse myeloma cell line NS0-derived recombinant human Pro-EGF Ser21-Arg1023 Accession # CAA28240
Immunogen affinity purified
RUO
Primary
1950
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only