missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro Coagulation Factor II/Thrombin Mouse anti-Human, Clone: 200710, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | Coagulation Factor II/Thrombin |
---|---|
Clone | 200710 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Pro Coagulation Factor II/Thrombin Ala44-Glu622 Accession # NP_000497 |
Gene ID (Entrez) | 2147 |
Gene Accession No. | NP_000497 |
Description
Coagulation Factor II/Thrombin Monoclonal specifically detects Coagulation Factor II/Thrombin in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Coagulation Factor II/Thrombin | |
Mouse myeloma cell line NS0-derived recombinant human Pro Coagulation Factor II/Thrombin Ala44-Glu622 Accession # NP_000497 | |
NP_000497 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Mouse | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
200710 | |
2147 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human Pro Coagulation Factor II/Thrombin in direct ELISAs and Western blots. It recognizes only Pro Coagulation Factor II/Thrombin, not active Thrombin. In Western blots, less than 5% cross-reactivity with recombinant human (rh) Factor X, rhKallikrein 1, 3, 5, and 11 is observed. | |
Unconjugated | |
Western Blot, Immunoprecipitation | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only