missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro BDNF Mouse anti-Human, Clone: 584421, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | BDNF |
---|---|
Clone | 584421 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Pro-BDNF Ala19-Arg247 Accession # P23560 |
Gene ID (Entrez) | 627 |
Gene Accession No. | P23560 |
Description
BDNF Monoclonal specifically detects BDNF in Human samples. It is validated for Immunohistochemistry.Specifications
BDNF | |
Chinese hamster ovary cell line CHO-derived recombinant human Pro-BDNF Ala19-Arg247 Accession # P23560 | |
P23560 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Mouse | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
584421 | |
627 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human Pro-BDNF in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) BDNF (mature), rh beta-NGF, rhNT-3, rhNT-4, rhPro-NT-3 or rhPro-NT-4 is observed. | |
Unconjugated | |
Immunohistochemistry | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only