missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRKD1 (PKC mu) Recombinant Human Protein
PRKC1 (PKC mu) is a serine/threonine protein kinase that is phospholipid-dependant and activated by diacylglycerol and phorbol ester TPA.
£1602.00 - £6900.00
Specifications
Accession Number | NP_002733 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 129.3kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15458543
|
Invitrogen™
PR7258C |
100μg |
£1602.00
100µg |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10587923
|
Invitrogen™
PV4041 |
1mg |
£6900.00
1mg |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PRKC1 behaves similarly to most PKC isoenzymes of the PKC family, however, PRKC1 differs in some structural and enzymatic features from the other PKC isotypes, and has been shown to be involved in the signal transduction of a wide range of biological responses.
Specifications
NP_002733 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
PKD Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
129.3kDa | |
RUO - research use only | |
PRKD1 (PKC mu), PKD | |
Human | |
GST Tag | |
Frozen |