missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRKCQ (PKC theta) Recombinant Human Protein
PRKCQ (PKC theta)
£297.00 - £6180.00
Specifications
Accession Number | NP_006248 |
---|---|
Gene Alias | PRKCQ (PKC theta) |
Species | Human |
Protein Tag | His Tag |
Expression System | Baculovirus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11529276
|
Invitrogen™
P2996 |
5μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15438393
|
Invitrogen™
PR4143B |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10719233
|
Invitrogen™
PV3605 |
1mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
The PKC family of serine⁄threonine kinases, including PRKCQ (PKC theta), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent.Specifications
NP_006248 | |
Human | |
Baculovirus |
PRKCQ (PKC theta) | |
His Tag |
For Research Use Only. Not for use in diagnostic procedures.