Learn More
Invitrogen™ PRKCH (PKC eta) Recombinant Human Protein
The PKC family of serine/threonine kinases, including PRKCH (PKC eta), is activated intracellularly by signal transduction pathways.
Brand: Invitrogen™ PR3588A
Code : NEW
Additional Details : Weight : 0.01000kg
Description
In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent.
Specifications
NP_006246 | |
82.2 kDa | |
PRKCH (PKC eta) | |
Human | |
His Tag | |
Ser⁄Thr Kinases: AGC Group | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |
Kinase Assay | |
100 μg | |
PKC Kinase Family | |
Recombinant | |
Storage Buffer: 20mM HEPES, pH 7.4, 250mM NaCl, 50% glycerol, 2mM EGTA, 2mM EDTA, 0.05% Triton X-100, 5mM DTT Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |