Learn More
Tocris Bioscience™ Pristimerin
Potent, reversible MAGL inhibitor
Brand:  Tocris Bioscience™ 373150
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13433514
  • £565.95 / 50mg
Estimated Shipment date
from Supplier 24-03-2020
Description
Description
Potent and reversible inhibitor of monoacylglycerol lipase (MGL) (IC50 = 93 nM). Also suppresses NF-κB activation via inhibition of proteasome chymotrypsin-like activity and IKKαβ. Displays antitumor, anti-inflammatory and antimicrobial activities.
Specifications
Specifications
Pristimerin
>99
50mg
Pristimerin
464.64