Learn More
Epredia™ PrintMate™ Biopsy Cassettes with Lids
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. PrintMate Cassettes provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
Brand: Epredia™ A84210080
Code : NEW
Additional Details : Weight : 3.00000kg
Description
1000 Biopsy Cassettes and 1000 LidsSpecifications
Small Square Pore | |
Aqua | |
Biopsy Cassette |