Learn More
Epredia™ PrintMate™ Embedding Cassettes with Lids, aqua
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. PrintMate Cassettes provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
Brand:  Epredia™ A84210071
Code : NEW
Additional Details : Weight : 3.00000kg
 View more versions of this product
Product Code. 15497624
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
Round Pore
Yes
Blue
Embedding Biopsy Cassette