Learn More
Epredia™ PrintMate™ AS Cassette Printer Accessories
Maintain the Epredia™ PrintMate™ Cassette Printer with PrintMate™ Accessories.
Brand:  Epredia™ A79510014
Code : 50
Additional Details : Weight : 8.74000kg
 View more versions of this product
Product Code. 12973745
  • £6630.00 / Each
Estimated Shipment: 16-05-2024
to see stock.
Description
Description
Accessories Include:
  • Collection system collection trays (box of 4)
  • Hopper
  • Hot foil tape, box of 5 (roll of tape prints approx. 20,000 cassettes)
Specifications
Specifications
Collection System
Collection system
PrintMate AS Cassette Printer, PrintMate Cassette Printer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products