missing translation for 'onlineSavingsMsg'
Learn More

PRIMA-1, Tocris Bioscience™

Restores mutant p53 activity; induces apoptosis

Brand:  Tocris (Bio-Techne) 186210

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13450144

  • £144.06 / 10mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selectively restores mutant p53 activity in tumor cells via activation of Bax and PUMA. Induces apoptosis and inhibits growth of tumors with mutant p53.
Specifications

Chemical Identifiers

5608-24-2
185.223
prima-1, 2,2-bis hydroxymethyl-1-azabicyclo 2,2,2, octan-3-one, 2,2-bis hydroxymethyl-1-azabicyclo 2.2.2 octan-3-one, 2,2-bis hydroxymethyl-3-quinuclidinone, p53 reactivation and induction of massive apoptosis, 2,2-bishydroxymethyl-1-aza-bicyclo 2.2.2 octan-3-one, 2,2-bis hydroxymethyl-1-azabicyclo 2,2,2 octan-3-one, prima 1, tocris-1862, prima 1;prima1
2,2-bis(hydroxymethyl)-1-azabicyclo[2.2.2]octan-3-one
C9H15NO3
RFBVBRVVOPAAFS-UHFFFAOYSA-N
322968
C1CN2CCC1C(=O)C2(CO)CO

Specifications

10mg
5608-24-2
prima-1, 2,2-bis hydroxymethyl-1-azabicyclo 2,2,2, octan-3-one, 2,2-bis hydroxymethyl-1-azabicyclo 2.2.2 octan-3-one, 2,2-bis hydroxymethyl-3-quinuclidinone, p53 reactivation and induction of massive apoptosis, 2,2-bishydroxymethyl-1-aza-bicyclo 2.2.2 octan-3-one, 2,2-bis hydroxymethyl-1-azabicyclo 2,2,2 octan-3-one, prima 1, tocris-1862, prima 1;prima1
C1CN2CCC1C(=O)C2(CO)CO
185.223
185.22
PRIMA-1
C9H15NO3
RFBVBRVVOPAAFS-UHFFFAOYSA-N
2,2-bis(hydroxymethyl)-1-azabicyclo[2.2.2]octan-3-one
322968
>99%
Videos
Documents